missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,6-Diaminoresorcinol Dihydrochloride 98.0+%, TCI America™
Supplier: TCI America D188825G
Specifications
| 4,6-Diaminoresorcinol Dihydrochloride | |
| C6H10Cl2N2O2 | |
| 25 g | |
| KUMOYHHELWKOCB-UHFFFAOYSA-N | |
| 4,6-diaminobenzene-1,3-diol;dihydrochloride | |
| 2733648 | |
| ≥98.0% (N,T) |
| 16523-31-2 | |
| MFCD00143239 | |
| 4,6-diaminoresorcinol dihydrochloride, 4,6-diaminobenzene-1,3-diol dihydrochloride, 4,6-diaminoresorcinol 2hcl, 1,3-benzenediol, 4,6-diamino-, dihydrochloride, 4,6-diamino resorcinol dihydrochloride, 4,6-diamino-benzene-1,3-diol 2hcl salt, 1,3-benzenediol, 4,6-diamino-, hydrochloride 1:2, 4,6-bis azanyl benzene-1,3-diol dihydrochloride, pubchem20873, acmc-1c78d | |
| C1=C(C(=CC(=C1N)O)O)N.Cl.Cl | |
| 213.058 | |
| 213.06 | |
| Crystalline Powder |
Chemical Identifiers
| 16523-31-2 | |
| 213.058 | |
| KUMOYHHELWKOCB-UHFFFAOYSA-N | |
| 2733648 | |
| C1=C(C(=CC(=C1N)O)O)N.Cl.Cl |
| C6H10Cl2N2O2 | |
| MFCD00143239 | |
| 4,6-diaminoresorcinol dihydrochloride, 4,6-diaminobenzene-1,3-diol dihydrochloride, 4,6-diaminoresorcinol 2hcl, 1,3-benzenediol, 4,6-diamino-, dihydrochloride, 4,6-diamino resorcinol dihydrochloride, 4,6-diamino-benzene-1,3-diol 2hcl salt, 1,3-benzenediol, 4,6-diamino-, hydrochloride 1:2, 4,6-bis azanyl benzene-1,3-diol dihydrochloride, pubchem20873, acmc-1c78d | |
| 4,6-diaminobenzene-1,3-diol;dihydrochloride |
Safety and Handling
EINECSNumber : (3)-4275
TSCA : Yes