missing translation for 'onlineSavingsMsg'
Learn More

4-[(6-Acryloyloxy)hexyloxy]-4'-cyanobiphenyl 98.0+%, TCI America™

Catalog No. A29861G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29861G 1 g
A29865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29861G Supplier TCI America Supplier No. A29861G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89823-23-4
Molecular Formula C22H23NO3
Molecular Weight (g/mol) 349.43
MDL Number MFCD16293619
InChI Key IGHSOWSFSFGPAZ-UHFFFAOYSA-N
PubChem CID 11416795
IUPAC Name 6-[4-(4-cyanophenyl)phenoxy]hexyl prop-2-enoate
SMILES C=CC(=O)OCCCCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 71°C
Color White-Yellow
Quantity 1 g
Synonym 4′C-[(6-Acryloyloxy)hexyloxy]-4-biphenylcarbonitrile, 6-[(4′C-Cyano-[1,1′C-biphenyl]-4-yl)oxy]hexyl Acrylate, Acrylic Acid 6-[(4′C-Cyano-[1,1′C-biphenyl]-4-yl)oxy]hexyl Ester
Formula Weight 349.43
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-[(6-Acryloyloxy)hexyloxy]-4′-cyanobiphenyl