missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethyl-1,2-phenylenediamine 98.0+%, TCI America™

Supplier:  TCI America D108325G

 View more versions of this product

Catalog No. D108325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4,5-Dimethyl-1,2-phenylenediamine
3171-45-7
Crystal-Powder at 20°C
MFCD00007729
XSZYBMMYQCYIPC-UHFFFAOYSA-N
4,5-dimethylbenzene-1,2-diamine
76635
≥98.0% (HPLC)
127°C
25 g
C8H12N2
4,5-dimethyl-1,2-phenylenediamine, 4,5-diamino-o-xylene, 4,5-dimethyl-o-phenylenediamine, 1,2-benzenediamine, 4,5-dimethyl, 1,2-diamino-4,5-dimethylbenzene, 4,5-dimethyl-1,2-benzenediamine, 4,5-dimethyl-benzene-1,2-diamine, 4,5-dimethyl-ortho-phenylenediamine, 4,5-dimethylphenylene-1,2-diamine, 4,5-dimethyl-o-phenylenediamine monohydrate
CC1=CC(N)=C(N)C=C1C
136.20
136.20

Chemical Identifiers

3171-45-7
136.20
XSZYBMMYQCYIPC-UHFFFAOYSA-N
76635
CC1=CC(N)=C(N)C=C1C
C8H12N2
MFCD00007729
4,5-dimethyl-1,2-phenylenediamine, 4,5-diamino-o-xylene, 4,5-dimethyl-o-phenylenediamine, 1,2-benzenediamine, 4,5-dimethyl, 1,2-diamino-4,5-dimethylbenzene, 4,5-dimethyl-1,2-benzenediamine, 4,5-dimethyl-benzene-1,2-diamine, 4,5-dimethyl-ortho-phenylenediamine, 4,5-dimethylphenylene-1,2-diamine, 4,5-dimethyl-o-phenylenediamine monohydrate
4,5-dimethylbenzene-1,2-diamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions