missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethyl-1,2-phenylenediamine 98.0+%, TCI America™

Catalog No. p-7126588
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10835G 5 g
D108325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D10835G Supplier TCI America Supplier No. D10835G

Chemical Identifiers

CAS 3171-45-7
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.20
MDL Number MFCD00007729
InChI Key XSZYBMMYQCYIPC-UHFFFAOYSA-N
Synonym 4,5-dimethyl-1,2-phenylenediamine, 4,5-diamino-o-xylene, 4,5-dimethyl-o-phenylenediamine, 1,2-benzenediamine, 4,5-dimethyl, 1,2-diamino-4,5-dimethylbenzene, 4,5-dimethyl-1,2-benzenediamine, 4,5-dimethyl-benzene-1,2-diamine, 4,5-dimethyl-ortho-phenylenediamine, 4,5-dimethylphenylene-1,2-diamine, 4,5-dimethyl-o-phenylenediamine monohydrate
PubChem CID 76635
IUPAC Name 4,5-dimethylbenzene-1,2-diamine
SMILES CC1=CC(N)=C(N)C=C1C

Specifications

Melting Point 127°C
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 136.20
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material 4,5-Dimethyl-1,2-phenylenediamine