missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrophenylacetic acid, 97%

Catalog No. AAA1888406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1888406 5 g
AAA1888403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1888406 Supplier Thermo Scientific Chemicals Supplier No. A1888406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 73357-18-3 | C10H11NO6 | 241.199 g/mol

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 73357-18-3
Molecular Formula C10H11NO6
Molecular Weight (g/mol) 241.199
MDL Number MFCD00118466
InChI Key WJPMJFUEMVXUCV-UHFFFAOYSA-N
Synonym 2-4,5-dimethoxy-2-nitrophenyl acetic acid, 4,5-dimethoxy-2-nitrophenyl acetic acid, 4,5-dimethoxy-2-nitrophenylacetic acid, 4,5-dimethoxy-2-nitro-phenyl-acetic acid, 4,5-dimethoxy-2-nitro-phenylacetic acid, benzeneacetic acid, 4,5-dimethoxy-2-nitro, maybridge1_002500, acmc-1bm2u, cbmicro_008990, cambridge id 5234450
PubChem CID 270941
IUPAC Name 2-(4,5-dimethoxy-2-nitrophenyl)acetic acid
SMILES COC1=C(C=C(C(=C1)CC(=O)O)[N+](=O)[O-])OC

Specifications

Melting Point 206°C to 208°C
Quantity 5 g
Formula Weight 241.2
Percent Purity 97%
Chemical Name or Material 4,5-Dimethoxy-2-nitrophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only