CAS RN 73357-18-3
IUPAC Name:
2-(4,5-dimethoxy-2-nitrophenyl)acetate
Synonyms:
2-(4,5-dimethoxy-2-nitrophenyl)acetate
Molecular Weight (g/mol):
240.19
Molecular Formula:
C10H10NO6
InChi Key:
WJPMJFUEMVXUCV-UHFFFAOYSA-M
SMILES:
COC1=CC(CC([O-])=O)=C(C=C1OC)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
4,5-Dimethoxy-2-nitrophenylacetic acid, 97%
4,5-Dimethoxy-2-nitrophenylacetic acid, 97%
CAS: 73357-18-3 Molecular Formula: C10H11NO6 Molecular Weight (g/mol): 241.199 MDL Number: MFCD00118466 InChI Key: WJPMJFUEMVXUCV-UHFFFAOYSA-N Synonym: 2-4,5-dimethoxy-2-nitrophenyl acetic acid,4,5-dimethoxy-2-nitrophenyl acetic acid,4,5-dimethoxy-2-nitrophenylacetic acid,4,5-dimethoxy-2-nitro-phenyl-acetic acid,4,5-dimethoxy-2-nitro-phenylacetic acid,benzeneacetic acid, 4,5-dimethoxy-2-nitro,maybridge1_002500,acmc-1bm2u,cbmicro_008990,cambridge id 5234450 PubChem CID: 270941 IUPAC Name: 2-(4,5-dimethoxy-2-nitrophenyl)acetic acid SMILES: COC1=C(C=C(C(=C1)CC(=O)O)[N+](=O)[O-])OC