missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrobenzonitrile 98.0+%, TCI America™

Catalog No. D29845G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29845G Supplier TCI America Supplier No. D29845G
Only null left

Chemical Identifiers

CAS 102714-71-6
Molecular Formula C9H8N2O4
Molecular Weight (g/mol) 208.173
InChI Key NQSQQGDTYKGCOT-UHFFFAOYSA-N
Synonym 1-Cyano-4,5-dimethoxy-2-nitrobenzene, 6-Nitroveratronitrile
PubChem CID 1511288
IUPAC Name 4,5-dimethoxy-2-nitrobenzonitrile
SMILES COC1=C(C=C(C(=C1)C#N)[N+](=O)[O-])OC

Specifications

Melting Point 167°C
Quantity 5 g
UN Number 3439
Formula Weight 208.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dimethoxy-2-nitrobenzonitrile