accessibility menu, dialog, popup

UserName

H3CONNOOOCH3

CAS RN 102714-71-6

IUPAC Name: 4,5-dimethoxy-2-nitrobenzonitrile
Synonyms: 4,5-dimethoxy-2-nitrobenzonitrile
Molecular Weight (g/mol): 208.17
Molecular Formula: C9H8N2O4
InChi Key: NQSQQGDTYKGCOT-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C(C(=C1)C#N)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results