Learn More
4,5-Dimethoxy-2-nitrobenzoic acid, 98%
CAS: 4998-07-6 | C9H8NO6 | 226.17 g/mol
$47.49 - $349.53
Chemical Identifiers
| CAS | 4998-07-6 |
|---|---|
| MDL Number | MFCD00014697 |
| IUPAC Name | 4-pyrrolidin-1-ylpiperidine |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0851006
|
Thermo Scientific Chemicals
L0851006 |
5 g |
N/A
|
|
|||||
|
AAL0851014
|
Thermo Scientific Chemicals
L0851014 |
25 g |
N/A
|
|
|||||
|
AAL0851022
|
Thermo Scientific Chemicals
L0851022 |
100 g |
N/A
|
|
|||||
Description
4,5-Dimethoxy-2-nitrobenzoic acid was used in the synthesis of 4,5-dimethoxy-2-nitrobenzamide and 6,7-dimethoxyquinazoline derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,5-Dimethoxy-2-nitrobenzoic acid was used in the synthesis of 4,5-dimethoxy-2-nitrobenzamide and 6,7-dimethoxyquinazoline derivatives.
Solubility
Insoluble in water. Soluble in DMSO and methanol. Solubility in hot 1mol/L NaOH (almost transparency).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 4998-07-6 | |
| MFCD00014697 | |
| 4-pyrrolidin-1-ylpiperidine |
Specifications
| 4998-07-6 | |
| C9H8NO6 | |
| 5 g | |
| 4,5-dimethoxy-2-nitrobenzoic acid, 6-nitroveratric acid, 3,4-dimethoxy-6-nitrobenzoic acid, benzoic acid, 4,5-dimethoxy-2-nitro, zlchem 681, acmc-2097gn, ksc496i5r, 4,5-dimethoxy-2-nitrobenzoicacid, 2-nitro-4,5-dimethoxybenzoic acid, 4,5-dimethoxy-2-nitro-benzoic acid | |
| WWCMFGBGMJAJRX-UHFFFAOYSA-M | |
| 226.17 | |
| 227.17 | |
| 4,5-Dimethoxy-2-nitrobenzoic acid |
| 192°C to 195°C | |
| MFCD00014697 | |
| 2123903 | |
| Insoluble in water. Soluble in DMSO and methanol. Solubility in hot 1mol/L NaOH (almost transparency). | |
| COC1=C(C=C(C(=C1)C(=O)O)[N+](=O)[O-])OC | |
| 78690 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 225-657-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only