missing translation for 'onlineSavingsMsg'
Learn More

4,5-Difluorophthalonitrile 95.0+%, TCI America™

Catalog No. p-7128020
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39631G 1 g
D39635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39631G Supplier TCI America Supplier No. D39631G

Chemical Identifiers

CAS 134450-56-9
Molecular Formula C8H2F2N2
Molecular Weight (g/mol) 164.115
InChI Key KNDUBEZZKOOYMJ-UHFFFAOYSA-N
Synonym 1,2-Dicyano-4,5-difluorobenzene
PubChem CID 11960962
IUPAC Name 4,5-difluorobenzene-1,2-dicarbonitrile
SMILES C1=C(C(=CC(=C1F)F)C#N)C#N

Specifications

Melting Point 114°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 164.11
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Difluorophthalonitrile