accessibility menu, dialog, popup

UserName

FFNN

CAS RN 134450-56-9

IUPAC Name: 4,5-difluorobenzene-1,2-dicarbonitrile
Synonyms: 4,5-difluorobenzene-1,2-dicarbonitrile
Molecular Weight (g/mol): 164.12
Molecular Formula: C8H2F2N2
InChi Key: KNDUBEZZKOOYMJ-UHFFFAOYSA-N
SMILES: FC1=C(F)C=C(C#N)C(=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results