missing translation for 'onlineSavingsMsg'
Learn More

4,5-Difluoro-2-hydroxybenzonitrile, 99%, Thermo Scientific Chemicals

Catalog No. p-7028749
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2615503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2615503 Supplier Thermo Scientific Chemicals Supplier No. H2615503

4,5-Difluoro-2-hydroxybenzonitrile crystals have been used as a practical means for monitoring, imaging, and other purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Difluoro-2-hydroxybenzonitrile crystals have been used as a practical means for monitoring, imaging, and other purposes.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 186590-36-3
Molecular Formula C7H3F2NO
Molecular Weight (g/mol) 155.10
MDL Number MFCD08275287
InChI Key ROLMZTIHUMKEAI-UHFFFAOYSA-N
Synonym 4,5-difluoro-2-hydroxy-benzonitrile, benzonitrile, 4,5-difluoro-2-hydroxy, acmc-1c7c6, 2-cyano-4,5-difluorophenol, 4,5-difluoro-2-hydroxybenzenecarbonitrile, benzonitrile, 4,5-difluoro-2-hydroxy-9ci
PubChem CID 23148985
IUPAC Name 4,5-difluoro-2-hydroxybenzonitrile
SMILES OC1=CC(F)=C(F)C=C1C#N

Specifications

Melting Point 161°C to 163°C
Quantity 1 g
UN Number UN3439
Formula Weight 155.1
Percent Purity 99%
Chemical Name or Material 4,5-Difluoro-2-hydroxybenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only