missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Dichloro-3(2H)-pyridazinone 98.0+%, TCI America™
Supplier: TCI America D2768100G
Specifications
| 4,5-Dichloro-3(2H)-pyridazinone | |
| 203°C | |
| C4H2Cl2N2O | |
| 100 g | |
| VJWXIRQLLGYIDI-UHFFFAOYSA-N | |
| 4,5-dichloro-2,3-dihydropyridazin-3-one | |
| 73247 | |
| ≥98.0% (GC,T) |
| 932-22-9 | |
| White | |
| MFCD00051504 | |
| 4,5-dichloro-3 2h-pyridazinone, 4,5-dichloropyridazin-3 2h-one, 3 2h-pyridazinone, 4,5-dichloro, 4,5-dichloropyridazin-3-ol, 4,5-dichloro-2h-pyridazin-3-one, 4,5-dichloro-3-hydroxypyridazine, 4,5-dichloro-2,3-dihydropyridazin-3-one, 4,5-dichloro-pyridazin-3-ol, 4,5-dichloro-3-pyridazinol | |
| ClC1=C(Cl)C(=O)NN=C1 | |
| 164.97 | |
| 164.97 | |
| Crystalline Powder |
Chemical Identifiers
| 932-22-9 | |
| 164.97 | |
| VJWXIRQLLGYIDI-UHFFFAOYSA-N | |
| 73247 | |
| ClC1=C(Cl)C(=O)NN=C1 |
| C4H2Cl2N2O | |
| MFCD00051504 | |
| 4,5-dichloro-3 2h-pyridazinone, 4,5-dichloropyridazin-3 2h-one, 3 2h-pyridazinone, 4,5-dichloro, 4,5-dichloropyridazin-3-ol, 4,5-dichloro-2h-pyridazin-3-one, 4,5-dichloro-3-hydroxypyridazine, 4,5-dichloro-2,3-dihydropyridazin-3-one, 4,5-dichloro-pyridazin-3-ol, 4,5-dichloro-3-pyridazinol | |
| 4,5-dichloro-2,3-dihydropyridazin-3-one |
Safety and Handling
RTECSNumber : UR6182000
TSCA : No