accessibility menu, dialog, popup

UserName

ClClONHN

CAS RN 932-22-9

IUPAC Name: 4,5-dichloro-2,3-dihydropyridazin-3-one
Synonyms: 4,5-dichloro-2H-pyridazin-3-one
Molecular Weight (g/mol): 164.97
Molecular Formula: C4H2Cl2N2O
InChi Key: VJWXIRQLLGYIDI-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C(=O)NN=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)