missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloro-3(2H)-pyridazinone 98.0+%, TCI America™

Catalog No. p-7127486
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D276825G 25 g
D2768100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D276825G Supplier TCI America Supplier No. D276825G
Only null left

Chemical Identifiers

CAS 932-22-9
Molecular Formula C4H2Cl2N2O
Molecular Weight (g/mol) 164.97
MDL Number MFCD00051504
InChI Key VJWXIRQLLGYIDI-UHFFFAOYSA-N
Synonym 4,5-dichloro-3 2h-pyridazinone, 4,5-dichloropyridazin-3 2h-one, 3 2h-pyridazinone, 4,5-dichloro, 4,5-dichloropyridazin-3-ol, 4,5-dichloro-2h-pyridazin-3-one, 4,5-dichloro-3-hydroxypyridazine, 4,5-dichloro-2,3-dihydropyridazin-3-one, 4,5-dichloro-pyridazin-3-ol, 4,5-dichloro-3-pyridazinol
PubChem CID 73247
IUPAC Name 4,5-dichloro-2,3-dihydropyridazin-3-one
SMILES ClC1=C(Cl)C(=O)NN=C1

Specifications

Melting Point 203°C
Color White
Quantity 25 g
Formula Weight 164.97
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dichloro-3(2H)-pyridazinone
RTECSNumber UR6182000
TSCA No