missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dibromoveratrole, 98+%

CAS: 37895-73-1 | C8H8Br2O2 | 295.958 g/mol

Supplier:  Thermo Scientific Chemicals L0199918

 View more versions of this product

Catalog No. AAL0199918


Only null left
Explore available promotions

Description

Description

4,5-Dibromoveratrole is used as chemical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Dibromoveratrole is used as chemical intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with Strong oxidizing agents.
Specifications

Specifications

4,5-Dibromoveratrole
90°C to 92°C
MFCD00014894
1953148
Insoluble in water.
COC1=CC(=C(C=C1OC)Br)Br
295.958
295.96
37895-73-1
C8H8Br2O2
50 g
4,5-dibromoveratrole, 4,5-dibromoveratrol, benzene, 1,2-dibromo-4,5-dimethoxy, acmc-1aeu5, amth147, 1.2-dibromo-4.5-dimethoxy-benzene, 1,2-bis bromanyl-4,5-dimethoxy-benzene
ZYCLQXMMFJREPJ-UHFFFAOYSA-N
1,2-dibromo-4,5-dimethoxybenzene
2758033
≥98%

Chemical Identifiers

37895-73-1
295.958
ZYCLQXMMFJREPJ-UHFFFAOYSA-N
2758033
COC1=CC(=C(C=C1OC)Br)Br
C8H8Br2O2
MFCD00014894
4,5-dibromoveratrole, 4,5-dibromoveratrol, benzene, 1,2-dibromo-4,5-dimethoxy, acmc-1aeu5, amth147, 1.2-dibromo-4.5-dimethoxy-benzene, 1,2-bis bromanyl-4,5-dimethoxy-benzene
1,2-dibromo-4,5-dimethoxybenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 253-705-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only