missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dibromoveratrole, 98+%

Catalog No. p-7043673
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0199909 10 g
AAL0199918 50 g
AAL0199930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0199909 Supplier Thermo Scientific Chemicals Supplier No. L0199909

CAS: 37895-73-1 | C8H8Br2O2 | 295.958 g/mol

4,5-Dibromoveratrole is used as chemical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Dibromoveratrole is used as chemical intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with Strong oxidizing agents.

Chemical Identifiers

CAS 37895-73-1
Molecular Formula C8H8Br2O2
Molecular Weight (g/mol) 295.958
MDL Number MFCD00014894
InChI Key ZYCLQXMMFJREPJ-UHFFFAOYSA-N
Synonym 4,5-dibromoveratrole, 4,5-dibromoveratrol, benzene, 1,2-dibromo-4,5-dimethoxy, acmc-1aeu5, amth147, 1.2-dibromo-4.5-dimethoxy-benzene, 1,2-bis bromanyl-4,5-dimethoxy-benzene
PubChem CID 2758033
IUPAC Name 1,2-dibromo-4,5-dimethoxybenzene
SMILES COC1=CC(=C(C=C1OC)Br)Br

Specifications

Melting Point 90°C to 92°C
Quantity 10 g
Beilstein 1953148
Solubility Information Insoluble in water.
Formula Weight 295.96
Percent Purity ≥98%
Chemical Name or Material 4,5-Dibromoveratrole
EINECSNumber 253-705-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only