missing translation for 'onlineSavingsMsg'
Learn More

4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene 98.0+%, TCI America™

Catalog No. p-7119731
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
25 g
5 g
Catalog No. Quantity
B27091G 1 g
B27095G 5 g
B270925G 25 g
3 options

Catalog No. B27091G

Supplier: TCI America B27091G

Only null left

Chemical Identifiers

CAS 161265-03-8
Molecular Formula C39H32OP2
Molecular Weight (g/mol) 578.63
MDL Number MFCD00233866
InChI Key CXNIUSPIQKWYAI-UHFFFAOYSA-N
Synonym xantphos, 4,5-bis diphenylphosphino-9,9-dimethylxanthene, 9,9-dimethyl-4,5-bis diphenylphosphino xanthene, 9,9-dimethyl-9h-xanthene-4,5-diyl bis diphenylphosphine, dimethylbisdiphenylphosphinoxanthene, unii-nmu72mog9b, nmu72mog9b, 9,9-dimethyl-9h-xanthene-4,5-diyl bis diphenylphosphane, 5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl-diphenylphosphane, 4,5-bis-diphenylphosphanyl-9,9-dimethyl-9h-xanthene
PubChem CID 636044
IUPAC Name [5-(diphenylphosphanyl)-9,9-dimethyl-9H-xanthen-4-yl]diphenylphosphane
SMILES CC1(C)C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 229°C
Color White-Yellow
Quantity 1 g
Formula Weight 578.63
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene