missing translation for 'onlineSavingsMsg'
Learn More

4,5-Bis(benzoylthio)-1,3-dithiole-2-thione 98.0+%, TCI America™

Catalog No. p-7118914
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11991G 1 g
B11995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11991G Supplier TCI America Supplier No. B11991G
Only null left

Chemical Identifiers

CAS 68494-08-6
Molecular Formula C17H10O2S5
Molecular Weight (g/mol) 406.565
MDL Number MFCD00059894
InChI Key YQZPSSPUVUGQQS-UHFFFAOYSA-N
PubChem CID 364673
IUPAC Name S-(5-benzoylsulfanyl-2-sulfanylidene-1,3-dithiol-4-yl) benzenecarbothioate
SMILES C1=CC=C(C=C1)C(=O)SC2=C(SC(=S)S2)SC(=O)C3=CC=CC=C3

Specifications

Melting Point 142°C
Quantity 1 g
Formula Weight 406.57
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Bis(benzoylthio)-1,3-dithiole-2-thione