Learn More
4',5,7-Trihydroxyisoflavone, 99+%
Inhibitor of tyrosine protein kinase | CAS: 446-72-0 | C15H10O5 | 270.24 g/mol
Supplier: Thermo Scientific Chemicals J6324103
Description
4',5,7-Trihydroxyisoflavone is used to inhibitor of tyrosine protein kinase. Genistein inhibits EGF-stimulated phosphorylation in cultured cells. Also used to explore signal transduction pathways.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4′,5,7-Trihydroxyisoflavone is used to inhibitor of tyrosine protein kinase. Genistein inhibits EGF-stimulated phosphorylation in cultured cells. Also used to explore signal transduction pathways.
Solubility
Soluble in chloroform, methanol. Insoluble in cold water.
Notes
Protect from heat. Store away from oxidizing agents.
Specifications
| 4',5,7-Trihydroxyisoflavone | |
| ∼300°C | |
| C15H10O5 | |
| 1 g | |
| 14,4391 | |
| Soluble in chloroform,methanol. Insoluble in cold water. | |
| OC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O | |
| 270.24 | |
| CHEBI:28088 | |
| ≥99% |
| 446-72-0 | |
| Brown to White | |
| MFCD00016952 | |
| 263823 | |
| genistein, prunetol, genisteol, 4',5,7-trihydroxyisoflavone, genisterin, sophoricol, 5,7,4'-trihydroxyisoflavone, bonistein, 5,7-dihydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, genestein | |
| TZBJGXHYKVUXJN-UHFFFAOYSA-N | |
| 5,7-dihydroxy-3-(4-hydroxyphenyl)chromen-4-one | |
| 5280961 | |
| 270.24 | |
| Powder |
Chemical Identifiers
| 446-72-0 | |
| 270.24 | |
| TZBJGXHYKVUXJN-UHFFFAOYSA-N | |
| 5280961 | |
| 5,7-dihydroxy-3-(4-hydroxyphenyl)chromen-4-one |
| C15H10O5 | |
| MFCD00016952 | |
| genistein, prunetol, genisteol, 4',5,7-trihydroxyisoflavone, genisterin, sophoricol, 5,7,4'-trihydroxyisoflavone, bonistein, 5,7-dihydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, genestein | |
| CHEBI:28088 | |
| OC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 207-174-9
RTECSNumber : NR2392000
TSCA : Yes
Recommended Storage : Store at -20°C
RUO – Research Use Only