missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5,6,7-Tetrahydrothieno[3,2-c]pyridine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America T277025G
Specifications
| 4,5,6,7-Tetrahydrothieno[3,2-c]pyridine Hydrochloride | |
| 228°C | |
| C7H10ClNS | |
| 25 g | |
| QMXCTKPNQFJZGK-UHFFFAOYSA-N | |
| 4,5,6,7-tetrahydrothieno[3,2-c]pyridine;hydrochloride | |
| 10176342 | |
| ≥98.0% (HPLC,N) |
| 28783-41-7 | |
| Yellow | |
| MFCD05861485 | |
| 4,5,6,7-tetrahydrothieno 3,2-c pyridine hydrochloride, 4,5,6,7-tetrahydrothieno 3,2,c pyridine hydrochloride, 4h,5h,6h,7h-thieno 3,2-c pyridine hydrochloride, 4,5,6,7-tetrahydrothieno 3,2-c pyridine hcl, 4,5,6,7-tetrahydrothieno 3,2,c pyridine hcl, unii-e0hb70dq7n, 4,5,6,7-tetrahydro-thieno 3,2-c pyridine hydrochloride, thieno 3,2-c pyridine, 4,5,6,7-tetrahydro-, hydrochloride | |
| C1CNCC2=C1SC=C2.Cl | |
| 175.674 | |
| 175.67 | |
| Crystalline Powder |
Chemical Identifiers
| 28783-41-7 | |
| 175.674 | |
| QMXCTKPNQFJZGK-UHFFFAOYSA-N | |
| 10176342 | |
| C1CNCC2=C1SC=C2.Cl |
| C7H10ClNS | |
| MFCD05861485 | |
| 4,5,6,7-tetrahydrothieno 3,2-c pyridine hydrochloride, 4,5,6,7-tetrahydrothieno 3,2,c pyridine hydrochloride, 4h,5h,6h,7h-thieno 3,2-c pyridine hydrochloride, 4,5,6,7-tetrahydrothieno 3,2-c pyridine hcl, 4,5,6,7-tetrahydrothieno 3,2,c pyridine hcl, unii-e0hb70dq7n, 4,5,6,7-tetrahydro-thieno 3,2-c pyridine hydrochloride, thieno 3,2-c pyridine, 4,5,6,7-tetrahydro-, hydrochloride | |
| 4,5,6,7-tetrahydrothieno[3,2-c]pyridine;hydrochloride |
Safety and Handling
TSCA : No