accessibility menu, dialog, popup

UserName

CAS RN 28783-41-7

IUPAC Name: 4H,5H,6H,7H-thieno[3,2-c]pyridine hydrochloride
Synonyms: 4H,5H,6H,7H-thieno[3,2-c]pyridine hydrochloride
Molecular Weight (g/mol): 175.67
Molecular Formula: C7H10ClNS
InChi Key: QMXCTKPNQFJZGK-UHFFFAOYSA-N
SMILES: Cl.C1CC2=C(CN1)C=CS2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results