missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydrothieno[3,2-c]pyridine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7137406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27705G 5 g
T277025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27705G Supplier TCI America Supplier No. T27705G

Chemical Identifiers

CAS 28783-41-7
Molecular Formula C7H10ClNS
Molecular Weight (g/mol) 175.674
MDL Number MFCD05861485
InChI Key QMXCTKPNQFJZGK-UHFFFAOYSA-N
Synonym 4,5,6,7-tetrahydrothieno 3,2-c pyridine hydrochloride, 4,5,6,7-tetrahydrothieno 3,2,c pyridine hydrochloride, 4h,5h,6h,7h-thieno 3,2-c pyridine hydrochloride, 4,5,6,7-tetrahydrothieno 3,2-c pyridine hcl, 4,5,6,7-tetrahydrothieno 3,2,c pyridine hcl, unii-e0hb70dq7n, 4,5,6,7-tetrahydro-thieno 3,2-c pyridine hydrochloride, thieno 3,2-c pyridine, 4,5,6,7-tetrahydro-, hydrochloride
PubChem CID 10176342
IUPAC Name 4,5,6,7-tetrahydrothieno[3,2-c]pyridine;hydrochloride
SMILES C1CNCC2=C1SC=C2.Cl

Specifications

Melting Point 228°C
Color Yellow
Quantity 5 g
Formula Weight 175.67
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,5,6,7-Tetrahydrothieno[3,2-c]pyridine Hydrochloride