missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Thiobis(6-tert-butyl-m-cresol) 98.0+%, TCI America™
Supplier: TCI America T0196100G
Specifications
| 4,4′-Thiobis(6-tert-butyl-m-cresol) | |
| 162°C | |
| C22H30O2S | |
| 100 g | |
| HXIQYSLFEXIOAV-UHFFFAOYSA-N | |
| 2-tert-butyl-4-[(5-tert-butyl-4-hydroxy-2-methylphenyl)sulfanyl]-5-methylphenol | |
| 7308 | |
| ≥98.0% (GC) |
| 96-69-5 | |
| White | |
| MFCD00026287 | |
| 4,4'-thiobis 6-tert-butyl-m-cresol, sumilizer wx, santonox bm, yoshinox sr, thioalkofen mbch, thioalkofen bmch, sumilizer wx-r, santowhite crystals, thioalkofen bm 4, disperse mb-61 | |
| CC1=CC(O)=C(C=C1SC1=CC(=C(O)C=C1C)C(C)(C)C)C(C)(C)C | |
| 358.54 | |
| 358.54 | |
| Crystalline Powder |
Chemical Identifiers
| 96-69-5 | |
| 358.54 | |
| HXIQYSLFEXIOAV-UHFFFAOYSA-N | |
| 7308 | |
| CC1=CC(O)=C(C=C1SC1=CC(=C(O)C=C1C)C(C)(C)C)C(C)(C)C |
| C22H30O2S | |
| MFCD00026287 | |
| 4,4'-thiobis 6-tert-butyl-m-cresol, sumilizer wx, santonox bm, yoshinox sr, thioalkofen mbch, thioalkofen bmch, sumilizer wx-r, santowhite crystals, thioalkofen bm 4, disperse mb-61 | |
| 2-tert-butyl-4-[(5-tert-butyl-4-hydroxy-2-methylphenyl)sulfanyl]-5-methylphenol |
Safety and Handling
EINECSNumber : (3)-1118
RTECSNumber : GP3150000
TSCA : Yes