accessibility menu, dialog, popup

UserName

CAS RN 96-69-5

CH3HOSOHH3CCH3CH3H3CCH3H3CCH3

CAS RN 96-69-5

IUPAC Name: 2-tert-butyl-4-[(5-tert-butyl-4-hydroxy-2-methylphenyl)sulfanyl]-5-methylphenol
Synonyms: 2-tert-butyl-4-[(5-tert-butyl-4-hydroxy-2-methylphenyl)sulfanyl]-5-methylphenol
Molecular Weight (g/mol): 358.54
Molecular Formula: C22H30O2S
InChi Key: HXIQYSLFEXIOAV-UHFFFAOYSA-N
SMILES: CC1=CC(O)=C(C=C1SC1=CC(=C(O)C=C1C)C(C)(C)C)C(C)(C)C
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results