missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Thiobis(6-tert-butyl-m-cresol) 98.0+%, TCI America™

Catalog No. T0196500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0196500G 500 g
T019625G 25 g
T0196100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0196500G Supplier TCI America Supplier No. T0196500G

Chemical Identifiers

CAS 96-69-5
Molecular Formula C22H30O2S
Molecular Weight (g/mol) 358.54
MDL Number MFCD00026287
InChI Key HXIQYSLFEXIOAV-UHFFFAOYSA-N
Synonym 4,4'-thiobis 6-tert-butyl-m-cresol, sumilizer wx, santonox bm, yoshinox sr, thioalkofen mbch, thioalkofen bmch, sumilizer wx-r, santowhite crystals, thioalkofen bm 4, disperse mb-61
PubChem CID 7308
IUPAC Name 2-tert-butyl-4-[(5-tert-butyl-4-hydroxy-2-methylphenyl)sulfanyl]-5-methylphenol
SMILES CC1=CC(O)=C(C=C1SC1=CC(=C(O)C=C1C)C(C)(C)C)C(C)(C)C

Specifications

Melting Point 162°C
Color White
Quantity 500 g
Formula Weight 358.54
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Thiobis(6-tert-butyl-m-cresol)
EINECSNumber (3)-1118
RTECSNumber GP3150000
TSCA Yes