missing translation for 'onlineSavingsMsg'
Learn More

4-(4-tert-Butylphenyl)benzoic Acid 98.0+%, TCI America™

Catalog No. B19865G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19865G 5 g
B19861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19865G Supplier TCI America Supplier No. B19865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5748-42-5
Molecular Formula C17H18O2
Molecular Weight (g/mol) 254.329
MDL Number MFCD01631300
InChI Key HZIOPONJCVOCFE-UHFFFAOYSA-N
PubChem CID 541479
IUPAC Name 4-(4-tert-butylphenyl)benzoic acid
SMILES CC(C)(C)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O

Specifications

Color White-Yellow
Quantity 5 g
Synonym 4-tert-Butylbiphenyl-4′-carboxylic Acid
Formula Weight 254.33
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-tert-Butylphenyl)benzoic Acid