accessibility menu, dialog, popup

UserName

CAS RN 5748-42-5

IUPAC Name: 4'-tert-butyl-[1,1'-biphenyl]-4-carboxylic acid
Synonyms: 4'-tert-butyl-[1,1'-biphenyl]-4-carboxylic acid
Molecular Weight (g/mol): 254.33
Molecular Formula: C17H18O2
InChi Key: HZIOPONJCVOCFE-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results