missing translation for 'onlineSavingsMsg'
Learn More

4-[4-(tert-Butoxycarbonyl)piperazino]benzoic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 162046-66-4
Molecular Formula C16H22N2O4
Molecular Weight (g/mol) 306.362
InChI Key BEDWYXZFIYMEJG-UHFFFAOYSA-N
Synonym 4-4-tert-butoxycarbonyl piperazin-1-yl benzoic acid, 4-4-tert-butoxycarbonyl piperazino benzoic acid, 4-4-carboxyphenyl piperazine-1-carboxylic acid tert-butyl ester, 4-4-boc-1-piperazineyl benzoic acid, 4-4-carboxy-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 1-4-carboxyphenyl-4-boc piperazine, 4-4-boc-piperazin-1-yl benzoic acid, 1-boc-4-4-carboxy-phenyl-piperazine, 1-4-boc-piperazin-1-yl-benzoic acid
PubChem CID 2795508
IUPAC Name 4-[4-[(2-methylpropan-2-yl)oxycarbonyl]piperazin-1-yl]benzoic acid
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC30001CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC30001DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC30001DE
View Documents
N/A 5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

162046-66-4
306.362
4-4-tert-butoxycarbonyl piperazin-1-yl benzoic acid, 4-4-tert-butoxycarbonyl piperazino benzoic acid, 4-4-carboxyphenyl piperazine-1-carboxylic acid tert-butyl ester, 4-4-boc-1-piperazineyl benzoic acid, 4-4-carboxy-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 1-4-carboxyphenyl-4-boc piperazine, 4-4-boc-piperazin-1-yl benzoic acid, 1-boc-4-4-carboxy-phenyl-piperazine, 1-4-boc-piperazin-1-yl-benzoic acid
4-[4-[(2-methylpropan-2-yl)oxycarbonyl]piperazin-1-yl]benzoic acid
C16H22N2O4
BEDWYXZFIYMEJG-UHFFFAOYSA-N
2795508
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)C(=O)O

Specifications

162046-66-4
C16H22N2O4
BEDWYXZFIYMEJG-UHFFFAOYSA-N
4-[4-[(2-methylpropan-2-yl)oxycarbonyl]piperazin-1-yl]benzoic acid
2795508
97%
97%
4-4-tert-butoxycarbonyl piperazin-1-yl benzoic acid, 4-4-tert-butoxycarbonyl piperazino benzoic acid, 4-4-carboxyphenyl piperazine-1-carboxylic acid tert-butyl ester, 4-4-boc-1-piperazineyl benzoic acid, 4-4-carboxy-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 1-4-carboxyphenyl-4-boc piperazine, 4-4-boc-piperazin-1-yl benzoic acid, 1-boc-4-4-carboxy-phenyl-piperazine, 1-4-boc-piperazin-1-yl-benzoic acid
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)C(=O)O
306.362
306.36
4-[4-(tert-Butoxycarbonyl)piperazino]benzoic acid
Videos
SDS
Documents

Documents