accessibility menu, dialog, popup

UserName

CH3H3CCH3OONNOO

CAS RN 162046-66-4

IUPAC Name: 4-{4-[(tert-butoxy)carbonyl]piperazin-1-yl}benzoate
Synonyms: 4-[4-(tert-butoxycarbonyl)piperazin-1-yl]benzoate
Molecular Weight (g/mol): 305.36
Molecular Formula: C16H21N2O4
InChi Key: BEDWYXZFIYMEJG-UHFFFAOYSA-M
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(C=C1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)