missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Nitrobenzyl)pyridine 98.0+%, TCI America™

Catalog No. p-7133667
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N01831G 1 g
N018325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N01831G Supplier TCI America Supplier No. N01831G
Only null left

Chemical Identifiers

CAS 1083-48-3
Molecular Formula C12H10N2O2
Molecular Weight (g/mol) 214.22
MDL Number MFCD00006445
InChI Key MNHKUCBXXMFQDM-UHFFFAOYSA-N
Synonym 4-4-nitrobenzyl pyridine, 4-4-nitrophenyl methyl pyridine, pyridine, 4-4-nitrophenyl methyl, pyridine, 4-p-nitrobenzyl, 1-p-nitrobenzyl pyridine, 4-p-nitrobenzyl pyridine, 4 4-nitrobenzyl primide, unii-0z6gs37n7h, gamma-p-nitrobenzyl pyridine
PubChem CID 14129
IUPAC Name 4-[(4-nitrophenyl)methyl]pyridine
SMILES [O-][N+](=O)C1=CC=C(CC2=CC=NC=C2)C=C1

Specifications

Melting Point 72°C
Color Yellow
Quantity 1 g
Formula Weight 214.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Nitrobenzyl)pyridine
RTECSNumber UT6360000
TSCA Yes