missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Nitrobenzyl)pyridine, 98+%

CAS: 1083-48-3 | C12H10N2O2 | 214.22 g/mol

Supplier:  Thermo Scientific Chemicals A1356406

 View more versions of this product

Catalog No. AAA1356406


Only null left
Explore available promotions

Description

Description

4-(4-Nitrobenzyl)pyridine is used in spectroscopic method for the determination of phosgene. It is also used in tests for alkylating agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Nitrobenzyl)pyridine is used in spectroscopic method for the determination of phosgene. It is also used in tests for alkylating agents.

Solubility
Soluble in acetone. Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong bases and strong acids.
Specifications

Specifications

4-(4-Nitrobenzyl)pyridine
69°C to 72°C
MFCD00006445
170877
4-4-nitrobenzyl pyridine, 4-4-nitrophenyl methyl pyridine, pyridine, 4-4-nitrophenyl methyl, pyridine, 4-p-nitrobenzyl, 1-p-nitrobenzyl pyridine, 4-p-nitrobenzyl pyridine, 4 4-nitrobenzyl primide, unii-0z6gs37n7h, gamma-p-nitrobenzyl pyridine
MNHKUCBXXMFQDM-UHFFFAOYSA-N
4-[(4-nitrophenyl)methyl]pyridine
14129
≥98%
1083-48-3
C12H10N2O2
5 g
Light sensitive
Soluble in acetone. Insoluble in water.
[O-][N+](=O)C1=CC=C(CC2=CC=NC=C2)C=C1
214.22
214.22

Chemical Identifiers

1083-48-3
214.22
MNHKUCBXXMFQDM-UHFFFAOYSA-N
14129
[O-][N+](=O)C1=CC=C(CC2=CC=NC=C2)C=C1
C12H10N2O2
MFCD00006445
4-4-nitrobenzyl pyridine, 4-4-nitrophenyl methyl pyridine, pyridine, 4-4-nitrophenyl methyl, pyridine, 4-p-nitrobenzyl, 1-p-nitrobenzyl pyridine, 4-p-nitrobenzyl pyridine, 4 4-nitrobenzyl primide, unii-0z6gs37n7h, gamma-p-nitrobenzyl pyridine
4-[(4-nitrophenyl)methyl]pyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c

H302-H315-H319-H335-H340

EINECSNumber : 214-108-2

RTECSNumber : UT6360000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only