missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(N,N-dimethylaniline), 98+%

Catalog No. p-7024178
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1846618 50 g
AAA1846630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1846618 Supplier Thermo Scientific Chemicals Supplier No. A1846618
Only null left

CAS: 101-61-1 | C17H22N2 | 254.377 g/mol

4,4'-Methylenebis(N,N-dimethylaniline)is used as a chemical intermediate in the manufacture of dyes and for its hydrochloric salt and is also used as an analytical reagent in the determination of lead.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Methylenebis(N,N-dimethylaniline)is used as a chemical intermediate in the manufacture of dyes and for its hydrochloric salt and is also used as an analytical reagent in the determination of lead.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 101-61-1
Molecular Formula C17H22N2
Molecular Weight (g/mol) 254.377
MDL Number MFCD00008317
InChI Key JNRLEMMIVRBKJE-UHFFFAOYSA-N
Synonym 4,4'-methylenebis n,n-dimethylaniline, tetra-base, tetrabase, michler's base, methane base, methylene base, michler's hydride, michler's methane, 4,4'-bis dimethylamino diphenylmethane, reduced michler's ketone
PubChem CID 7567
ChEBI CHEBI:34370
IUPAC Name 4-[[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)CC2=CC=C(C=C2)N(C)C

Specifications

Melting Point 85°C to 88°C
Boiling Point 155°C to 157°C (0.1 mmHg)
Flash Point 178°C (352°F)
Quantity 50 g
UN Number UN3077
Beilstein 479956
Merck Index 14,6176
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 254.38
Percent Purity ≥98%
Chemical Name or Material 4,4'-Methylenebis(N,N-dimethylaniline)
Show More Show Less
Hazard Category H350
Hazard Statement GHS H Statement
H350
May cause cancer.
Precautionary Statement P201-P202-P281-P308+P313-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 202-959-2
RTECSNumber BY2500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only