missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(N,N-dimethylaniline), 98%

CAS: 101-61-1 | C17H22N2 | 254.37 g/mol

$64.16 - $64.16

Chemical Identifiers

CAS 101-61-1
Molecular Formula C17H22N2
Molecular Weight (g/mol) 254.37
MDL Number MFCD00008317
InChI Key JNRLEMMIVRBKJE-UHFFFAOYSA-N
Synonym 4,4'-methylenebis n,n-dimethylaniline, tetra-base, tetrabase, michler's base, methane base, methylene base, michler's hydride, michler's methane, 4,4'-bis dimethylamino diphenylmethane, reduced michler's ketone
PubChem CID 7567
ChEBI CHEBI:34370
IUPAC Name 4-[[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)CC2=CC=C(C=C2)N(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC126740050
SDS View Documents
Thermo Scientific Chemicals
126740050
5 g Glass bottle
Each for $64.16
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

101-61-1
254.37
JNRLEMMIVRBKJE-UHFFFAOYSA-N
7567
4-[[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline
C17H22N2
MFCD00008317
4,4'-methylenebis n,n-dimethylaniline, tetra-base, tetrabase, michler's base, methane base, methylene base, michler's hydride, michler's methane, 4,4'-bis dimethylamino diphenylmethane, reduced michler's ketone
CHEBI:34370
CN(C)C1=CC=C(C=C1)CC2=CC=C(C=C2)N(C)C

Specifications

101-61-1
100.0
Beige-Brown or Gray or White
178°C
98%
C17H22N2
MFCD00008317
13, 239
4,4'-methylenebis n,n-dimethylaniline, tetra-base, tetrabase, michler's base, methane base, methylene base, michler's hydride, michler's methane, 4,4'-bis dimethylamino diphenylmethane, reduced michler's ketone
JNRLEMMIVRBKJE-UHFFFAOYSA-N
4-[[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline
7567
254.37
Crystalline Powder, Crystals or Chunks
99.0
86.0°C to 90.0°C
390.0°C
Authentic
Glass bottle
H2NC6H4CH2C6H4NH2
5 g
15, 6255
Solubility in water: insoluble. Other solubilities: soluble in benzene,ether,carbon disulfide and,acids,slightly soluble in cold alcohol,more soluble in,hot alcohol
CN(C)C1=CC=C(C=C1)CC2=CC=C(C=C2)N(C)C
254.37
CHEBI:34370
98%
4, 4'-Methylenebis(N, N-dimethylaniline), 99+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause cancer.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-959-2

RTECSNumber : BY5250000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only