missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(benzenesulfonyl Chloride) 95.0+%, TCI America™

Catalog No. M15561G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M15561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M15561G Supplier TCI America Supplier No. M15561G

Chemical Identifiers

CAS 3119-64-0
Molecular Formula C13H10Cl2O4S2
Molecular Weight (g/mol) 365.24
MDL Number MFCD00012363
InChI Key YKMMQCFZCFAHOU-UHFFFAOYSA-N
Synonym Bis(4-chlorosulfonylphenyl)methane
PubChem CID 3368486
IUPAC Name 4-{[4-(chlorosulfonyl)phenyl]methyl}benzene-1-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(CC2=CC=C(C=C2)S(Cl)(=O)=O)C=C1

Specifications

Melting Point 125°C
Color White
Quantity 1 g
UN Number 3261
Formula Weight 365.24
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Methylenebis(benzenesulfonyl Chloride)