CAS RN 3119-64-0
IUPAC Name:
4-{[4-(chlorosulfonyl)phenyl]methyl}benzene-1-sulfonyl chloride
Synonyms:
4-{[4-(chlorosulfonyl)phenyl]methyl}benzenesulfonyl chloride
Molecular Weight (g/mol):
365.24
Molecular Formula:
C13H10Cl2O4S2
InChi Key:
YKMMQCFZCFAHOU-UHFFFAOYSA-N
SMILES:
ClS(=O)(=O)C1=CC=C(CC2=CC=C(C=C2)S(Cl)(=O)=O)C=C1
1
–
1
of
1
results
1
4,4'-Methylenebis(benzenesulfonyl Chloride) 95.0+%, TCI America™
CAS: 3119-64-0 Molecular Formula: C13H10Cl2O4S2 Molecular Weight (g/mol): 365.24 MDL Number: MFCD00012363 InChI Key: YKMMQCFZCFAHOU-UHFFFAOYSA-N Synonym: Bis(4-chlorosulfonylphenyl)methane PubChem CID: 3368486 IUPAC Name: 4-{[4-(chlorosulfonyl)phenyl]methyl}benzene-1-sulfonyl chloride SMILES: ClS(=O)(=O)C1=CC=C(CC2=CC=C(C=C2)S(Cl)(=O)=O)C=C1