missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(2,6-dimethylphenol) 98.0+%, TCI America™

Catalog No. p-7132538
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M109925G 25 g
M1099500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M109925G Supplier TCI America Supplier No. M109925G
Only null left

Chemical Identifiers

CAS 5384-21-4
Molecular Formula C17H20O2
Molecular Weight (g/mol) 256.345
MDL Number MFCD00055640
InChI Key AZZWZMUXHALBCQ-UHFFFAOYSA-N
PubChem CID 79345
IUPAC Name 4-[(4-hydroxy-3,5-dimethylphenyl)methyl]-2,6-dimethylphenol
SMILES CC1=CC(=CC(=C1O)C)CC2=CC(=C(C(=C2)C)O)C

Specifications

Melting Point 176°C
Color White-Yellow
Quantity 25 g
Synonym Bis(4-hydroxy-3,5-dimethylphenyl)methane, 4,4′C-Dihydroxy-3,3′C,5,5′C-tetramethyldiphenylmethane, 2,2′C,6,6′C-Tetramethyl-4,4′C-methylenediphenol
Formula Weight 256.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Methylenebis(2,6-dimethylphenol)
EINECSNumber (4)-1632
TSCA No