accessibility menu, dialog, popup

UserName

CH3CH3OHH3CH3CHO

CAS RN 5384-21-4

IUPAC Name: 4-[(4-hydroxy-3,5-dimethylphenyl)methyl]-2,6-dimethylphenol
Synonyms: 4-[(4-hydroxy-3,5-dimethylphenyl)methyl]-2,6-dimethylphenol
Molecular Weight (g/mol): 256.35
Molecular Formula: C17H20O2
InChi Key: AZZWZMUXHALBCQ-UHFFFAOYSA-N
SMILES: CC1=CC(CC2=CC(C)=C(O)C(C)=C2)=CC(C)=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results