missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Methylenebis(2,6-diethylaniline) 98.0+%, TCI America™
$74.56 - $490.25
Chemical Identifiers
| CAS | 13680-35-8 |
|---|---|
| Molecular Formula | C21H30N2 |
| Molecular Weight (g/mol) | 310.485 |
| MDL Number | MFCD00071552 |
| InChI Key | NWIVYGKSHSJHEF-UHFFFAOYSA-N |
| Synonym | 4,4'-methylenebis 2,6-diethylaniline, benzenamine, 4,4'-methylenebis 2,6-diethyl, bis 4-amino-3,5-diethylphenyl methane, 4,4'-methylene-bis-2,6-diethylaniline, 4-4-amino-3,5-diethylphenyl methyl-2,6-diethylaniline, 4,4/'-methylenebis 2,6-diethylaniline, 4-4-amino-3,5-diethylphenyl methyl-2,6-diethylphenylamine, lonzacure r m-dea, acmc-1bxlo |
| PubChem CID | 83656 |
| IUPAC Name | 4-[(4-amino-3,5-diethylphenyl)methyl]-2,6-diethylaniline |
| SMILES | CCC1=CC(=CC(=C1N)CC)CC2=CC(=C(C(=C2)CC)N)CC |
Chemical Identifiers
| 13680-35-8 | |
| 310.485 | |
| NWIVYGKSHSJHEF-UHFFFAOYSA-N | |
| 83656 | |
| CCC1=CC(=CC(=C1N)CC)CC2=CC(=C(C(=C2)CC)N)CC |
| C21H30N2 | |
| MFCD00071552 | |
| 4,4'-methylenebis 2,6-diethylaniline, benzenamine, 4,4'-methylenebis 2,6-diethyl, bis 4-amino-3,5-diethylphenyl methane, 4,4'-methylene-bis-2,6-diethylaniline, 4-4-amino-3,5-diethylphenyl methyl-2,6-diethylaniline, 4,4/'-methylenebis 2,6-diethylaniline, 4-4-amino-3,5-diethylphenyl methyl-2,6-diethylphenylamine, lonzacure r m-dea, acmc-1bxlo | |
| 4-[(4-amino-3,5-diethylphenyl)methyl]-2,6-diethylaniline |
Specifications
| 13680-35-8 | |
| 205°C | |
| MFCD00071552 | |
| 4,4'-methylenebis 2,6-diethylaniline, benzenamine, 4,4'-methylenebis 2,6-diethyl, bis 4-amino-3,5-diethylphenyl methane, 4,4'-methylene-bis-2,6-diethylaniline, 4-4-amino-3,5-diethylphenyl methyl-2,6-diethylaniline, 4,4/'-methylenebis 2,6-diethylaniline, 4-4-amino-3,5-diethylphenyl methyl-2,6-diethylphenylamine, lonzacure r m-dea, acmc-1bxlo | |
| CCC1=CC(=CC(=C1N)CC)CC2=CC(=C(C(=C2)CC)N)CC | |
| 310.485 | |
| 310.49 | |
| Crystalline Powder |
| 90°C | |
| C21H30N2 | |
| 25 g | |
| NWIVYGKSHSJHEF-UHFFFAOYSA-N | |
| 4-[(4-amino-3,5-diethylphenyl)methyl]-2,6-diethylaniline | |
| 83656 | |
| ≥98.0% (GC,T) | |
| 4,4′-Methylenebis(2,6-diethylaniline) |
Safety and Handling
EINECSNumber : (4)-1614
TSCA : Yes