CAS RN 13680-35-8
CAS RN 13680-35-8
IUPAC Name:
4-[(4-amino-3,5-diethylphenyl)methyl]-2,6-diethylaniline
Synonyms:
4-[(4-amino-3,5-diethylphenyl)methyl]-2,6-diethylaniline
Molecular Weight (g/mol):
310.49
Molecular Formula:
C21H30N2
InChi Key:
NWIVYGKSHSJHEF-UHFFFAOYSA-N
SMILES:
CCC1=CC(CC2=CC(CC)=C(N)C(CC)=C2)=CC(CC)=C1N
1
–
1
of
1
results
1
4,4'-Methylenebis(2,6-diethylaniline) 98.0+%, TCI America™
CAS: 13680-35-8 Molecular Formula: C21H30N2 Molecular Weight (g/mol): 310.485 MDL Number: MFCD00071552 InChI Key: NWIVYGKSHSJHEF-UHFFFAOYSA-N Synonym: 4,4'-methylenebis 2,6-diethylaniline,benzenamine, 4,4'-methylenebis 2,6-diethyl,bis 4-amino-3,5-diethylphenyl methane,4,4'-methylene-bis-2,6-diethylaniline,4-4-amino-3,5-diethylphenyl methyl-2,6-diethylaniline,4,4/'-methylenebis 2,6-diethylaniline,4-4-amino-3,5-diethylphenyl methyl-2,6-diethylphenylamine,lonzacure r m-dea,acmc-1bxlo PubChem CID: 83656 IUPAC Name: 4-[(4-amino-3,5-diethylphenyl)methyl]-2,6-diethylaniline SMILES: CCC1=CC(=CC(=C1N)CC)CC2=CC(=C(C(=C2)CC)N)CC