missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(2,6-diethylaniline) 98.0+%, TCI America™

Catalog No. M189725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M189725G 25 g
M1897500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M189725G Supplier TCI America Supplier No. M189725G
Only null left

Chemical Identifiers

CAS 13680-35-8
Molecular Formula C21H30N2
Molecular Weight (g/mol) 310.485
MDL Number MFCD00071552
InChI Key NWIVYGKSHSJHEF-UHFFFAOYSA-N
Synonym 4,4'-methylenebis 2,6-diethylaniline, benzenamine, 4,4'-methylenebis 2,6-diethyl, bis 4-amino-3,5-diethylphenyl methane, 4,4'-methylene-bis-2,6-diethylaniline, 4-4-amino-3,5-diethylphenyl methyl-2,6-diethylaniline, 4,4/'-methylenebis 2,6-diethylaniline, 4-4-amino-3,5-diethylphenyl methyl-2,6-diethylphenylamine, lonzacure r m-dea, acmc-1bxlo
PubChem CID 83656
IUPAC Name 4-[(4-amino-3,5-diethylphenyl)methyl]-2,6-diethylaniline
SMILES CCC1=CC(=CC(=C1N)CC)CC2=CC(=C(C(=C2)CC)N)CC

Specifications

Melting Point 90°C
Boiling Point 205°C
Quantity 25 g
Formula Weight 310.49
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Methylenebis(2,6-diethylaniline)
EINECSNumber (4)-1614
TSCA Yes