missing translation for 'onlineSavingsMsg'
Learn More

4-[(4-Methyl-1-piperazinyl)methyl]benzoic Acid Dihydrochloride 98.0+%, TCI America™

Catalog No. M21105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21105G 5 g
M211025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21105G Supplier TCI America Supplier No. M21105G

Chemical Identifiers

CAS 106261-49-8
Molecular Formula C13H20Cl2N2O2
Molecular Weight (g/mol) 307.215
MDL Number MFCD07772867
InChI Key ISHROKOWRJDOSN-UHFFFAOYSA-N
Synonym 4-4-methylpiperazin-1-yl methyl benzoic acid dihydrochloride, 4-4-methyl-1-piperazinyl methyl benzoic acid dihydrochloride, 4-4-methylpiperazin-1-yl methyl, 4-4-methylpiperazinomethyl benzoic acid dihydrochloride, benzoic acid, 4-4-methyl-1-piperazinyl methyl-, dihydrochloride, zlchem 396, ksc909c2h, 4-4-methyl piperazino methyl benzoic acid 2hcl, 4-4-methylpiperazin-1-yl methyl benzoic acid dih
PubChem CID 10335346
IUPAC Name 4-[(4-methylpiperazin-1-yl)methyl]benzoic acid;dihydrochloride
SMILES CN1CCN(CC1)CC2=CC=C(C=C2)C(=O)O.Cl.Cl

Specifications

Color White
Quantity 5 g
Formula Weight 307.22
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-[(4-Methyl-1-piperazinyl)methyl]benzoic Acid Dihydrochloride