missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Isopropylidenediphenoxyacetic Acid 98.0+%, TCI America™

Catalog No. I03565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03565G 5 g
I035625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03565G Supplier TCI America Supplier No. I03565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3539-42-2
Molecular Formula C19H20O6
Molecular Weight (g/mol) 344.36
MDL Number MFCD00060114
InChI Key ZYGPJSLWIIMUMO-UHFFFAOYSA-N
Synonym Bisphenol A O,O-Diacetic Acid
PubChem CID 12359775
IUPAC Name 2-(4-{2-[4-(carboxymethoxy)phenyl]propan-2-yl}phenoxy)acetic acid
SMILES CC(C)(C1=CC=C(OCC(O)=O)C=C1)C1=CC=C(OCC(O)=O)C=C1

Specifications

Melting Point 179°C
Color White-Yellow
Quantity 5 g
Formula Weight 344.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Isopropylidenediphenoxyacetic Acid