missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Hydroxyphenyl)-2-butanone, 98%

Catalog No. AAA1406414
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1406414 25 g
AAA1406422 100 g
AAA1406430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1406414 Supplier Thermo Scientific Chemicals Supplier No. A1406414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5471-51-2 | C10H12O2 | 164.204 g/mol

4-(4-Hydroxyphenyl)-2-butanone is used in perfumery, in cosmetics, and as a food additive to impart a fruity odor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Hydroxyphenyl)-2-butanone is used in perfumery, in cosmetics, and as a food additive to impart a fruity odor.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 5471-51-2
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00002394
InChI Key NJGBTKGETPDVIK-UHFFFAOYSA-N
Synonym 4-4-hydroxyphenyl-2-butanone, raspberry ketone, 4-4-hydroxyphenyl butan-2-one, frambinone, oxyphenalon, rheosmin, 4-p-hydroxyphenyl-2-butanone, 4-hydroxybenzylacetone, betuligenol, rasketone
PubChem CID 21648
ChEBI CHEBI:68656
IUPAC Name 4-(4-hydroxyphenyl)butan-2-one
SMILES CC(=O)CCC1=CC=C(C=C1)O

Specifications

Melting Point 80°C to 84°C
Quantity 25 g
Beilstein 776080
Solubility Information Insoluble in water.
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material 4-(4-Hydroxyphenyl)-2-butanone
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 226-806-4
RTECSNumber EL8925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only