missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Hydroxy-3-methoxyphenyl)-2-butanone 98.0+%, TCI America™

Catalog No. p-7130975
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H131425G 25 g
H1314100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H131425G Supplier TCI America Supplier No. H131425G
Only null left

Chemical Identifiers

CAS 122-48-5
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00048232
InChI Key OJYLAHXKWMRDGS-UHFFFAOYSA-N
Synonym zingerone, vanillylacetone, zingiberone, 4-4-hydroxy-3-methoxyphenyl butan-2-one, zingherone, 4-4-hydroxy-3-methoxyphenyl-2-butanone, gingerone, vanillyl acetone, 0-paradol, 2-butanone, 4-4-hydroxy-3-methoxyphenyl
PubChem CID 31211
ChEBI CHEBI:68657
IUPAC Name 4-(4-hydroxy-3-methoxyphenyl)butan-2-one
SMILES CC(=O)CCC1=CC(=C(C=C1)O)OC

Specifications

Melting Point 41°C
Color White-Yellow
Quantity 25 g
Formula Weight 194.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Hydroxy-3-methoxyphenyl)-2-butanone
RTECSNumber EL8900000
TSCA Yes
Recommended Storage Refrigerator