missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Hydroxy-3-methoxyphenyl)-2-butanone, 97%

Catalog No. p-7028036
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2515003 1 g
AAB2515006 5 g
AAB2515014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2515003 Supplier Thermo Scientific Chemicals Supplier No. B2515003
Only null left

CAS: 122-48-5 | C11H14O3 | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122-48-5
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00048232
InChI Key OJYLAHXKWMRDGS-UHFFFAOYSA-N
Synonym zingerone, vanillylacetone, zingiberone, 4-4-hydroxy-3-methoxyphenyl butan-2-one, zingherone, 4-4-hydroxy-3-methoxyphenyl-2-butanone, gingerone, vanillyl acetone, 0-paradol, 2-butanone, 4-4-hydroxy-3-methoxyphenyl
PubChem CID 31211
ChEBI CHEBI:68657
IUPAC Name 4-(4-hydroxy-3-methoxyphenyl)butan-2-one
SMILES CC(=O)CCC1=CC(=C(C=C1)O)OC

Specifications

Melting Point 39°C to 42°C
Quantity 1 g
Formula Weight 194.23
Percent Purity 97%
Chemical Name or Material 4-(4-Hydroxy-3-methoxyphenyl)-2-butanone
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 204-548-3
RTECSNumber EL8900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only