missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Diphenyl-2-cyclohexen-1-one 98.0+%, TCI America™
$233.00 - $766.13
Chemical Identifiers
| CAS | 4528-64-7 |
|---|---|
| Molecular Formula | C18H16O |
| Molecular Weight (g/mol) | 248.325 |
| MDL Number | MFCD00075159 |
| InChI Key | LUFGFXHHSGISSL-UHFFFAOYSA-N |
| PubChem CID | 138280 |
| IUPAC Name | 4,4-diphenylcyclohex-2-en-1-one |
| SMILES | C1CC(C=CC1=O)(C2=CC=CC=C2)C3=CC=CC=C3 |
Chemical Identifiers
| 4528-64-7 | |
| 248.325 | |
| LUFGFXHHSGISSL-UHFFFAOYSA-N | |
| 4,4-diphenylcyclohex-2-en-1-one |
| C18H16O | |
| MFCD00075159 | |
| 138280 | |
| C1CC(C=CC1=O)(C2=CC=CC=C2)C3=CC=CC=C3 |
Specifications
| 4528-64-7 | |
| White-Yellow | |
| MFCD00075159 | |
| LUFGFXHHSGISSL-UHFFFAOYSA-N | |
| 4,4-diphenylcyclohex-2-en-1-one | |
| 138280 | |
| ≥98.0% (GC) | |
| 4,4-Diphenyl-2-cyclohexen-1-one |
| 98°C | |
| C18H16O | |
| 1 g | |
| C1CC(C=CC1=O)(C2=CC=CC=C2)C3=CC=CC=C3 | |
| 248.325 | |
| 248.33 | |
| Crystalline Powder |
Safety and Handling
TSCA : No