accessibility menu, dialog, popup

UserName

CAS RN 4528-64-7

O

CAS RN 4528-64-7

IUPAC Name: 4,4-diphenylcyclohex-2-en-1-one
Synonyms: 4,4-diphenylcyclohex-2-en-1-one
Molecular Weight (g/mol): 248.33
Molecular Formula: C18H16O
InChi Key: LUFGFXHHSGISSL-UHFFFAOYSA-N
SMILES: O=C1CCC(C=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results