missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dinitrodiphenyl Ether 99.0+%, TCI America™
Supplier: TCI America D08321G
Specifications
| 4,4′-Dinitrodiphenyl Ether | |
| 144°C | |
| C12H8N2O5 | |
| 1 g | |
| MWAGUKZCDDRDCS-UHFFFAOYSA-N | |
| 1-nitro-4-(4-nitrophenoxy)benzene | |
| 7568 | |
| ≥99.0% (GC) |
| 101-63-3 | |
| Yellow | |
| MFCD00059182 | |
| 4,4′C-Dinitrodiphenyl Oxide | |
| C1=CC(=CC=C1[N+](=O)[O-])OC2=CC=C(C=C2)[N+](=O)[O-] | |
| 260.205 | |
| 260.21 | |
| Crystalline Powder |
Chemical Identifiers
| 101-63-3 | |
| 260.205 | |
| MWAGUKZCDDRDCS-UHFFFAOYSA-N | |
| 1-nitro-4-(4-nitrophenoxy)benzene |
| C12H8N2O5 | |
| MFCD00059182 | |
| 7568 | |
| C1=CC(=CC=C1[N+](=O)[O-])OC2=CC=C(C=C2)[N+](=O)[O-] |
Safety and Handling
EINECSNumber : (3)-0883
RTECSNumber : KN3450000
TSCA : Yes