missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dinitrobiphenyl 99.0+%, TCI America™

Supplier:  TCI America D08265G

Catalog No. D08265G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4,4′-Dinitrobiphenyl
242°C
C12H8N2O4
5 g
C1=CC(=CC=C1C2=CC=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]
244.206
244.21
Crystalline Powder
1528-74-1
Yellow
MFCD00039744
BDLNCFCZHNKBGI-UHFFFAOYSA-N
1-nitro-4-(4-nitrophenyl)benzene
15216
≥99.0% (GC)

Chemical Identifiers

1528-74-1
244.206
BDLNCFCZHNKBGI-UHFFFAOYSA-N
1-nitro-4-(4-nitrophenyl)benzene
C12H8N2O4
MFCD00039744
15216
C1=CC(=CC=C1C2=CC=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : DV4000000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions